ENA311759566 Enamine

3-ethoxypropanimidamide hydrochloride

  • MDL number MFCD09260544
  • Cas Number : 1170847-92-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70608

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70608
Availability details for
Quantity
  • Availability
  • Add to Cart