ENA311759567 Enamine

2-methoxy-2-phenylpropanoic acid

  • MDL number MFCD00231151
  • Cas Number : 7472-67-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-70609

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70609
Availability details for
Quantity
  • Availability
  • Add to Cart