ENA311759568 Enamine

[3-fluoro-4-(propan-2-yloxy)phenyl]methanamine

  • MDL number MFCD11191825
  • Cas Number : 1039337-06-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70610

Safety Information

msds https://www.enaminestore.com/catalog/EN300-70610
Availability details for
Quantity
  • Availability
  • Add to Cart