ENA311759571 Enamine

2-[(2-chlorophenyl)sulfanyl]-2-phenylacetic acid

  • MDL number MFCD09715740
  • Cas Number : 938267-73-3
  • Molecular Weight : 278.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70613

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70613
Availability details for
Quantity
  • Availability
  • Add to Cart