ENA311759577 Enamine

1-(propan-2-yl)-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD08704773
  • Cas Number : 1041644-48-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 106...108
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43703

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-43703
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart