ENA311759581 Enamine

2-[(2-methyl-2,3-dihydro-1H-indol-1-yl)methyl]furan-3-carboxylic acid

  • MDL number MFCD12580125
  • Cas Number : 1156139-50-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-70624

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70624
Availability details for
Quantity
  • Availability
  • Add to Cart