ENA311759584 Enamine

3-(chloromethyl)-5-(oxan-4-yl)-1,2,4-oxadiazole

  • MDL number MFCD17242175
  • Cas Number : 1249078-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63665

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63665
Availability details for
Quantity
  • Availability
  • Add to Cart