ENA311759593 Enamine

1,3-diethyl 2-(2-methylpropanamido)propanedioate

  • MDL number MFCD12644195
  • Cas Number : 153180-28-0
  • Molecular Weight : 245.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-43887

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43887
Availability details for
Quantity
  • Availability
  • Add to Cart