ENA311759594 Enamine

2-{3-[(4-chlorophenyl)methoxy]phenyl}acetic acid

  • MDL number MFCD16385749
  • Cas Number : 916908-15-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70647

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70647
Availability details for
Quantity
  • Availability
  • Add to Cart