ENA311759595 Enamine

2-{3-[(3-methylphenyl)methoxy]phenyl}acetic acid

  • MDL number MFCD16385746
  • Cas Number : 1258649-85-2
  • Molecular Weight : 256.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70648

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70648
Availability details for
Quantity
  • Availability
  • Add to Cart