ENA311759596 Enamine

2-{3-[(2-chlorophenyl)methoxy]phenyl}acetic acid

  • MDL number MFCD16385751
  • Cas Number : 1258650-20-2
  • Molecular Weight : 276.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70649

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70649
Availability details for
Quantity
  • Availability
  • Add to Cart