ENA311759611 Enamine

ethyl 2-(methylamino)benzoate

  • MDL number MFCD11174593
  • Cas Number : 35472-56-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-70665

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70665
Availability details for
Quantity
  • Availability
  • Add to Cart