ENA311759617 Enamine

3-(propan-2-yl)-1-(pyrimidin-2-yl)-1H-pyrazol-5-amine

  • MDL number MFCD12192737
  • Cas Number : 1153082-81-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-70683

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70683
Availability details for
Quantity
  • Availability
  • Add to Cart