ENA311759622 Enamine

[2-(4-methylphenoxy)phenyl]methanamine

  • MDL number MFCD07365250
  • Cas Number : 289718-11-2
  • Molecular Weight : 213.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70688

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70688
Availability details for
Quantity
  • Availability
  • Add to Cart