ENA311759625 Enamine

2-[(2-methylphenyl)sulfanyl]-2-phenylacetic acid

  • MDL number MFCD09719943
  • Cas Number : 18626-45-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100...102
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70693

Safety Information

msds https://www.enaminestore.com/catalog/EN300-70693
Availability details for
Quantity
  • Availability
  • Add to Cart