ENA311759627 Enamine

2-{[1-(2,2,2-trifluoroethyl)-1H-1,2,3,4-tetrazol-5-yl]sulfanyl}acetic acid

  • MDL number MFCD11202805
  • Cas Number : 1038303-87-5
  • Molecular Weight : 242.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-64619

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64619
Availability details for
Quantity
  • Availability
  • Add to Cart