ENA311759630 Enamine

1-(piperidin-2-yl)ethan-1-ol

  • MDL number MFCD17976979
  • Cas Number : 54160-32-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 35-40
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64867

Safety Information

signalword Danger
pcodes P260,P264,P270,P280,P301+P312,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P330,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-64867
s_phrases_eu 26-27-28-36/37/39-45
pictograms GHS05,GHS07
hstatements H302,H314
Availability details for
Quantity
  • Availability
  • Add to Cart