ENA311759632 Enamine

tert-butyl N-(1-cyanopropyl)carbamate

  • MDL number MFCD07366974
  • Cas Number : 657424-07-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-70700

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70700
Availability details for
Quantity
  • Availability
  • Add to Cart