ENA311759636 Enamine

5-phenyl-1,2-oxazole-3-carboxamide

  • MDL number MFCD00464227
  • Cas Number : 23088-52-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 202...204
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70704

Safety Information

msds https://www.enaminestore.com/catalog/EN300-70704
Availability details for
Quantity
  • Availability
  • Add to Cart