ENA311759637 Enamine

3-methoxythiophene-2-carboxamide

  • MDL number MFCD01680278
  • Cas Number : 78031-17-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70705

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70705
Availability details for
Quantity
  • Availability
  • Add to Cart