ENA311759643 Enamine

3-chloro-4-cyanobenzene-1-sulfonamide

  • MDL number MFCD11212967
  • Cas Number : 1094691-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 157...159
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70715

Safety Information

msds https://www.enaminestore.com/catalog/EN300-70715
Availability details for
Quantity
  • Availability
  • Add to Cart