ENA311759654 Enamine

methyl 6-oxo-2-(pyridin-3-yl)-1,6-dihydropyrimidine-5-carboxylate

  • MDL number MFCD17977110
  • Molecular Weight : 231.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70730

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70730
Availability details for
Quantity
  • Availability
  • Add to Cart