ENA311759656 Enamine

4-ethyl-6-hydrazinyl-2-(pyridin-2-yl)pyrimidine

  • MDL number MFCD16727038
  • Cas Number : 1249703-49-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-70741

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70741
Availability details for
Quantity
  • Availability
  • Add to Cart