ENA311759658 Enamine

4-methyl-1-phenyl-1H-pyrazol-3-amine

  • MDL number MFCD16845430
  • Cas Number : 66000-37-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-70743

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70743
Availability details for
Quantity
  • Availability
  • Add to Cart