ENA311759661 Enamine

5-(2-phenylpropan-2-yl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD12104390
  • Cas Number : 1032463-93-6
  • Molecular Weight : 219.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70747

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70747
Availability details for
Quantity
  • Availability
  • Add to Cart