ENA311759664 Enamine

[2-(2-methylphenoxy)phenyl]methanamine

  • MDL number MFCD07365248
  • Cas Number : 870121-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-70750

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70750
Availability details for
Quantity
  • Availability
  • Add to Cart