ENA311759670 Enamine

2-[(1,1-dioxo-1lambda6-thiolan-3-yl)carbamoyl]cyclohexane-1-carboxylic acid

  • MDL number MFCD09907041
  • Cas Number : 1053161-05-9
  • Molecular Weight : 289.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70762

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70762
Availability details for
Quantity
  • Availability
  • Add to Cart