ENA311759674 Enamine

N-(2-methoxyethyl)-1H-1,3-benzodiazol-2-amine

  • MDL number MFCD09034748
  • Cas Number : 924867-97-0
  • Molecular Weight : 191.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70766

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70766
Availability details for
Quantity
  • Availability
  • Add to Cart