ENA311759680 Enamine

N-[3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)phenyl]acetamide

  • MDL number MFCD02223124
  • Cas Number : 61862-29-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185...187
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44333

Safety Information

msds https://www.enaminestore.com/catalog/EN300-44333
Availability details for
Quantity
  • Availability
  • Add to Cart