ENA311759683 Enamine

1H,2H,4H-pyrido[2,3-d][1,3]oxazine-2,4-dione

  • MDL number MFCD09863994
  • Cas Number : 21038-63-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 207...209
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70781

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-70781
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart