ENA311759685 Enamine

(2-phenyl-1,3-thiazol-5-yl)methanol

  • MDL number MFCD09694149
  • Cas Number : 859485-91-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 72...74
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70786

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-70786
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart