ENA311759688 Enamine

5-(3-methylthiophen-2-yl)-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD10690986
  • Cas Number : 1092308-05-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70789

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70789
Availability details for
Quantity
  • Availability
  • Add to Cart