ENA311759690 Enamine

(2S)-1-(prop-2-yn-1-yl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD09757462
  • Molecular Weight : 153.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2S)-1-(2-propynyl)-2-pyrrolidinecarboxylic acid
Inchi Code 1S/C8H11NO2/c1-2-5-9-6-3-4-7(9)8(10)11/h1,7H,3-6H2,(H,10,11)/t7-/m0/s1
Inchi Key YOBBRBOMLUMFMP-ZETCQYMHSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70791

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70791
Availability details for
Quantity
  • Availability
  • Add to Cart