ENA311759691 Enamine

(2,5-dimethylphenyl)methanethiol

  • MDL number MFCD09807604
  • Cas Number : 22182-98-5
  • Molecular Weight : 152.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70792

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70792
Availability details for
Quantity
  • Availability
  • Add to Cart