ENA311759699 Enamine

2-[(2,5-difluorophenyl)sulfanyl]-2-phenylacetic acid

  • MDL number MFCD12569684
  • Cas Number : 1097125-22-8
  • Molecular Weight : 280.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70800

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70800
Availability details for
Quantity
  • Availability
  • Add to Cart