ENA311759702 Enamine

1-cyclopentylpyrrolidin-3-amine

  • MDL number MFCD11640875
  • Cas Number : 933713-03-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-70803

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70803
Availability details for
Quantity
  • Availability
  • Add to Cart