ENA311759707 Enamine

2-chloro-1-[2-(4-methoxyphenyl)pyrrolidin-1-yl]ethan-1-one

  • MDL number MFCD13154469
  • Cas Number : 1184158-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-46094

Safety Information

msds https://www.enaminestore.com/catalog/EN300-46094
Availability details for
Quantity
  • Availability
  • Add to Cart