ENA311759710 Enamine

ethyl 2-(5-bromo-2-methoxyphenyl)-2-oxoacetate

  • MDL number MFCD16061579
  • Cas Number : 1191261-77-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70812

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70812
Availability details for
Quantity
  • Availability
  • Add to Cart