ENA311759733 Enamine

methyl({[2-(prop-2-yn-1-yloxy)phenyl]methyl})amine hydrochloride

  • MDL number MFCD17078404
  • Cas Number : 1258640-56-0
  • Molecular Weight : 211.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-70843

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70843
Availability details for
Quantity
  • Availability
  • Add to Cart