ENA311759735 Enamine

3-{[2-(aminomethyl)phenyl]methyl}-1,3-diazaspiro[4.5]decane-2,4-dione

  • MDL number MFCD09813223
  • Cas Number : 1016763-30-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70845

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70845
Availability details for
Quantity
  • Availability
  • Add to Cart