ENA311759743 Enamine

1-(3-bromophenyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD12134476
  • Cas Number : 1156994-32-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-71260

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71260
Availability details for
Quantity
  • Availability
  • Add to Cart