ENA311759745 Enamine

2-(pyridin-2-yl)-3H,4H,5H,6H,7H-cyclopenta[d]pyrimidin-4-one

  • MDL number MFCD14653757
  • Cas Number : 874779-29-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71262

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71262
Availability details for
Quantity
  • Availability
  • Add to Cart