ENA311759753 Enamine

1-cyclopentanecarbonylpyrrolidine-2-carboxylic acid

  • MDL number MFCD00522584
  • Cas Number : 1417985-23-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125...127
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71273

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71273
Availability details for
Quantity
  • Availability
  • Add to Cart