ENA311759758 Enamine

(prop-2-yn-1-yl)[(pyridin-3-yl)methyl]amine

  • MDL number MFCD07411961
  • Cas Number : 128813-42-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-71281

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71281
Availability details for
Quantity
  • Availability
  • Add to Cart