ENA311759761 Enamine

N-(6-amino-1,3-benzothiazol-2-yl)-2-morpholin-4-ylacetamide

  • MDL number MFCD09735582
  • Cas Number : 953736-70-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 228...230
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71284

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71284
Availability details for
Quantity
  • Availability
  • Add to Cart