ENA311759765 Enamine

3-methylpyridin-4-amine

  • MDL number MFCD01704431
  • Cas Number : 1990-90-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67367

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-67367
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart