ENA311759768 Enamine

3-[(2-oxooxolan-3-yl)sulfamoyl]benzoic acid

  • MDL number MFCD13271523
  • Cas Number : 1105058-09-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-71295

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71295
Availability details for
Quantity
  • Availability
  • Add to Cart