ENA311759771 Enamine

(cyclohexylmethyl)[(pyridin-4-yl)methyl]amine

  • MDL number MFCD09688717
  • Cas Number : 931992-77-7
  • Molecular Weight : 204.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-71298

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71298
Availability details for
Quantity
  • Availability
  • Add to Cart