ENA311759772 Enamine

1-(5-bromo-2-fluorophenyl)-2,2,2-trifluoroethan-1-ol

  • MDL number MFCD16067891
  • Cas Number : 1039827-16-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-71299

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71299
Availability details for
Quantity
  • Availability
  • Add to Cart