ENA311759778 Enamine

5-(2-aminophenyl)-5-methylimidazolidine-2,4-dione

  • MDL number MFCD12190038
  • Cas Number : 858204-20-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-68254

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68254
Availability details for
Quantity
  • Availability
  • Add to Cart